Learn More
tert-Butyl Methyl Ether-d12, CDN
Product Code. 30320957
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320957
Brand: CDN CDND53140.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl-tert-Butyl Ether D12
CAS 284487-64-5
Molecular Formula C5D12O
Quantity 0.25 g
Synonym 1,1,1,3,3,3-Hexadeuterio-2-(trideuteriomethoxy)-2-(trideuteriomethyl)propane, tert-Butyl methyl ether-d12
InChI Formula InChI=1 S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3,4D3
SMILES [2 H]C([2 H])([2 H])OC(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 2-(methoxy-d3)-2-(methyl-d3)propane-1,1,1,3,3,3-d6
Molecular Weight (g/mol) 100.22
Formula Weight 100.16 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity