missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium n-Octyl-d17 Sulfate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Sodium Octyl-D17 Sulfate |
| CAS | 1219802-16-0 |
| Molecular Formula | C8 D17 O4 S . Na |
| Quantity | 0.25 g |
| Synonym | Sodium n-octyl-d17 sulfate, Sulfuric acid monooctyl-d17 ester sodium salt, Sodium octyl-D17 sulfate, Sodium octyl sulfate D17 (octyl-D17), Sodium caprylyl-d17 sulfate |
| InChI Formula | InChI=1 S/C8H18O4S.Na/c1-2-3-4-5-6-7-8-12-13(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OS(=O)(=O)[O-] |
| IUPAC Name | sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecadeuteriooctyl sulfate |
| Molecular Weight (g/mol) | 249.378 |
| Formula Weight | 249.181 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction