Learn More
Pyrrolidine-2,2,5,5-d4, CDN
Product Code. 30315573
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315573
Brand: CDN CDND59460.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Pyrrolidine-2,2,5,5-D4
CAS 42403-25-8
Molecular Formula C4H5D4N
Quantity 0.25 g
Synonym 2,2,5,5-Tetradeuteropyrrolidine
InChI Formula InChI=1 S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2/i3D2,4D2
SMILES [2 H]C1([2 H])CCC([2 H])([2 H])N1
IUPAC Name pyrrolidine-2,2,5,5-d4
Molecular Weight (g/mol) 75.15
Formula Weight 75.1 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity