missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxy-d5-acetic Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Phenoxy-d5-acetic Acid |
| CAS | 154492-74-7 |
| Molecular Formula | C8 2H5 H3 O3 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C8H8O3/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5 H,6H2,(H,9,10)/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(OCC(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 2-(2,3,4,5,6-pentadeuteriophenoxy)acetic acid |
| Molecular Weight (g/mol) | 157.18 |
| Formula Weight | 157.0787 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction