missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentanoic-5,5,5-d3 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Pentanoic-5,5,5-d3 Acid |
| CAS | 83741-76-8 |
| Molecular Formula | C5 D3 H7 O2 |
| Quantity | 0.25 g |
| Synonym | Pentanoic-5,5,5-d3 acid (9 CI), delta;-Trideuteropentanoic acid, Pentanoic acid-5,5,5-d3, Valeric acid-d3, Valeric-delta;,delta;,delta;-d3 acid, Pentanoic-5,5,5-D3 acid |
| InChI Formula | InChI=1 S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCC(=O)O |
| IUPAC Name | 5,5,5-trideuteriopentanoic acid |
| Molecular Weight (g/mol) | 105.15 |
| Formula Weight | 105.087 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction