Learn More
Pentanedioic-d6 Anhydride, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Pentanedioic-D6 Anhydride |
| CAS | 1219794-53-2 |
| Molecular Formula | C5 D6 O3 |
| Quantity | 0.25 g |
| Synonym | Dihydro-2 H-pyran-2,6(3 H)-dione-3,3,4,4,5,5-d6, Glutaric anhydride-d6, Glutaric-d6 acid anhydride, Oxacyclohexane-2,6-dione-d6, Pentanedioic-2,2,3,3,4,4-d6 acid anhydride, Glutaric-d6 anhydride |
| InChI Formula | InChI=1 S/C5H6O3/c6-4-2-1-3-5(7)8-4/h1-3H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C1([2 H])C(=O)OC(=O)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 3,3,4,4,5,5-hexadeuteriooxane-2,6-dione |
| Molecular Weight (g/mol) | 120.1363 |
| Formula Weight | 120.0694 g/mol |
| Show More |