Learn More
p-Toluene-2,3,5,6-d4-sulfonamide, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Tolyl-d4-sulfonamide |
| CAS | 1219795-34-2 |
| Molecular Formula | C7 2H4 H5 N O2 S |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5 H,1H3,(H2,8,9,10)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1C)S(=O)(=O)N |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-methylbenzenesulfonamide |
| Molecular Weight (g/mol) | 175.24 |
| Formula Weight | 175.0605 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |