missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl-1,1,3,3,3-d5 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Propanol-1,1,3,3,3-D5 |
| CAS | 188894-71-5 |
| Molecular Formula | C3 D5 H3 O |
| Quantity | 0.25 g |
| Synonym | 1-Propan-1,1,3,3,3-d5-ol (9 CI), 1,1,3,3,3-Pentadeuteropropanol, 1,1,3,3,3-Pentadeuteropropan-1-ol, Propyl-1,1,3,3,3-d5 alcohol, 1-Propanol-1,1,3,3,3-D5 |
| InChI Formula | InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])CC([2 H])([2 H])O |
| IUPAC Name | 1,1,3,3,3-pentadeuteriopropan-1-ol |
| Molecular Weight (g/mol) | 65.126 |
| Formula Weight | 65.089 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction