missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentadecyl-d31 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Pentadecanol-D31 |
| CAS | 349553-87-3 |
| Molecular Formula | C15 D31 H O |
| Quantity | 0.25 g |
| Synonym | 1-Pentadecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-d31-ol (9 CI), 1-Pentadecanol-D31, n-Pentadecanol-d31, Pentadecyl-d31 alcohol, n-Pentadecyl-d31 alcohol |
| InChI Formula | InChI=1 S/C15H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h16H,2-15H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,15-hentriacontadeuteriopentadecan-1-ol |
| Molecular Weight (g/mol) | 259.605 |
| Formula Weight | 259.44 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction