Learn More
n-Nonylbenzene-2,3,4,5,6-d5, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | n-Nonylbenzene-2,3,4,5,6-D5 |
| CAS | 20216-93-7 |
| Molecular Formula | C15 D5 H19 |
| Quantity | 0.25 g |
| Synonym | Nonane, 1-(phenyl-d5)- (8 CI), n-Nonylbenzene-2,3,4,5,6-D5, Nonylbenzene-d5, Nonylbenzene-d5 (phenyl-d5), 1-(Phenyl-d5)nonane |
| InChI Formula | InChI=1 S/C15H24/c1-2-3-4-5-6-7-9-12-15-13-10-8-11-14-15/h8,10-11,13-14 H,2-7,9,12H2,1H3/i8D,10 D,11 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CCCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-nonylbenzene |
| Molecular Weight (g/mol) | 209.382 |
| Formula Weight | 209.219 g/mol |
| Show More |