missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-n-Dodecyl-d25-N,N-dimethyl-3-ammonio-1-propanesulfonate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 3-(N-Dodecyl-N,N-dimethylammonio)-1-propanesulfonate-D25 (N-dodecyl-D25) |
| Molecular Formula | C17 D25 H12 N O3 S |
| Quantity | 0.25 g |
| Synonym | N,N-Dimethyl-N-(3-sulfopropyl)-1-dodecanaminium-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25 inner salt, 3-(Dodecyl-d25-dimethylammonio)-1-propanesulfonic acid, 3-(N-Dodecyl-N,N-dimethylammonio)-1-propanesulfonate-D25 (N-dodecyl-D25), N-n-Dodecyl-d25-N,N-dimethyl-3-ammonio-1-propanesulfonate |
| InChI Formula | InChI=1 S/C17H37NO3S/c1-4-5-6-7-8-9-10-11-12-13-15-18(2,3)16-14-17-22(19,20)21/h4-17H2,1-3H3/i1D3,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,15D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C)(C)CCCS(=O)(=O)[O-] |
| IUPAC Name | 3-[dimethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl)azaniumyl]propane-1-sulfonate |
| Molecular Weight (g/mol) | 360.7 |
| Formula Weight | 360.406 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction