Learn More
n-Hexyl-d13 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1-Hexanol-D13 |
| CAS | 204244-84-8 |
| Molecular Formula | C6 D13 H O |
| Quantity | 0.25 g |
| Synonym | 1-Hexan-1,1,2,2,3,3,4,4,5,5,6,6,6-d13-ol (9 CI), 1-Hexanol-d13, n-Hexyl-d13 alcohol, 1-Hexanol-1,1,2,2,3,3,4,4,5,5,6,6,6-d13 |
| InChI Formula | InChI=1 S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecadeuteriohexan-1-ol |
| Molecular Weight (g/mol) | 115.2549 |
| Formula Weight | 115.1861 g/mol |
| Show More |