Learn More
n-Hexadecyl-16,16,16-d3 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Hexadecanol-16,16,16-D3 |
| CAS | 75736-52-6 |
| Molecular Formula | C16 D3 H31 O |
| Quantity | 0.25 g |
| Synonym | 1-Hexadecan-16,16,16-d3-ol (9 CI, ACI), [16,16,16-2H3]Hexadecanol, n-Hexadecanol-16,16,16-d3, n-Hexadecyl-16,16,16-d3 alcohol, Cetyl alcohol-ω-d3, Cetyl-16,16,16-d3 alcohol, 1-Hexadecanol-16,16,16-D3 |
| InChI Formula | InChI=1 S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCO |
| IUPAC Name | 16,16,16-trideuteriohexadecan-1-ol |
| Molecular Weight (g/mol) | 245.459 |
| Formula Weight | 245.28 g/mol |
| Show More |