Learn More
n-Heptyl-5,5,6,6,7,7,7-d7 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1-Heptanol-5,5,6,6,7,7,7-D7 |
| CAS | 1219804-99-5 |
| Molecular Formula | C7 D7 H9 O |
| Quantity | 0.25 g |
| Synonym | n-Heptyl-5,5,6,6,7,7,7-d7 alcohol, 1-Heptan-5,5,6,6,7,7,7-d7-ol, Heptyl-5,5,6,6,7,7,7-d7 alcohol, 1-Heptanol-5,5,6,6,7,7,7-D7 |
| InChI Formula | InChI=1 S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCO |
| IUPAC Name | 5,5,6,6,7,7,7-heptadeuterioheptan-1-ol |
| Molecular Weight (g/mol) | 123.245 |
| Formula Weight | 123.164 g/mol |
| Show More |