missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-alanine-2,3,3,3-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-D-alanine-2,3,3,3-d4 |
| CAS | 2708343-30-8 |
| Molecular Formula | C5 D4 H5 N O3 |
| Quantity | 0.25 g |
| Synonym | N-Acetyl-D-alanine-2,3,3,3-D4, N-Acetyl-D-alanine-d4, (2 R)-2-Acetamido-2,3,3,3-tetradeuteriopropanoic acid, (R)-2-Acetamidopropionic-2,3,3,3-d4 acid |
| InChI Formula | InChI=1 S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/t3-/m1/s1/i1D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])[C@@]([2 H])(NC(=O)C)C(=O)O |
| IUPAC Name | (2 R)-2-acetamido-2,3,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 135.155 |
| Formula Weight | 135.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction