Learn More
L-Phenylalanine-3,3-d2, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | L-Phenylalanine-D2 (propanoic-3,3-D2 acid) |
| CAS | 221346-31-2 |
| Molecular Formula | C9 D2 H9 N O2 |
| Quantity | 0.25 g |
| Synonym | L-Phenylalanine-β,β-d2, Phenylalanine-β,β-d2, (2 S)-2-Amino-3,3-dideuterio-3-phenylpropanoic acid, L-Phenylalanine-3,3-d2, L-Phenylalanine-d2, (2 S)-2-Amino-3-phenylpropanoic-3,3-d2 acid, (2 S)-2-Amino-3-phenylpropanoic-3,3-d2 acid, (2 S)-2-Amino-3-phenylpropanoic-3,3-d2 acid |
| InChI Formula | InChI=1 S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8 H,6,10H2,(H,11,12)/t8-/m0/s1/i6D2 |
| SMILES | [2 H]C([2 H])([C@H](N)C(=O)O)c1ccccc1 |
| IUPAC Name | (2 S)-2-amino-3,3-dideuterio-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 167.2015 |
| Formula Weight | 167.0915 g/mol |
| Show More |