Learn More
L-Phenyl-d5-alanine-N-FMOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine-2,3,4,5,6-D5 |
| CAS | 225918-67-2 |
| Molecular Formula | C24 D5 H16 N O4 |
| Quantity | 0.25 g |
| Synonym | L-Phenyl-d5-alanine, N-[(9 H-fluoren-9-ylmethoxy)carbonyl]- (9 CI), N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine-2,3,4,5,6-D5, (2 S)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-3-(phenyl-d5)propanoic acid, L-Phenyl-d5-alanine-N-FMOC, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-L-phenylalanine-d5 (phenyl-d5), N-(9-Fluorenylmethoxycarbonyl)-L-phenyl-d5-alanine, N-Fmoc-L-phenylalanine-d5 |
| InChI Formula | InChI=1 S/C24H21NO4/c26-23(27)22(14-16-8-2-1-3-9-16)25-24(28)29-15-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h1-13,21-22 H,14-15H2,(H,25,28)(H,26,27)/t22-/m0/s1/i1D,2 D,3 D,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C[C@H](NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | (2 S)-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 392.4587 |
| Formula Weight | 392.1784 g/mol |
| Show More |