Learn More
L-Isoleucine-2-d1, CDN
Product Code. 30318451
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30318451
Brand: CDN CDND63530.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Isoleucine-2-d1
CAS 127290-60-2
Molecular Formula C6 D H12 N O2
Quantity 0.25 g
Synonym L-Isoleucine-2-d1, L-Isoleucine-2-d (9 CI, ACI), L-Isoleucine-2-D, (2 S,3 S)-2-Amino-3-methylpentanoic-2-d acid
InChI Formula InChI=1 S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1/i5D
SMILES [2 H][C@](N)([C@@H](C)CC)C(=O)O
IUPAC Name (2 S,3 S)-2-amino-2-deuterio-3-methylpentanoic acid
Molecular Weight (g/mol) 132.179
Formula Weight 132.101 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity