Learn More
L-Alanine-2,3,3,3-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Alanine d4 |
| CAS | 18806-29-6 |
| Molecular Formula | C3 2H4 H3 N O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1/i1D3,2 D |
| SMILES | [2 H]C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 93.12 |
| Formula Weight | 93.0728 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |