Learn More
Hexanedioic-d8 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Hexanedioic Acid-D8 |
| CAS | 52089-65-3 |
| Molecular Formula | C6 D8 H2 O4 |
| Quantity | 0.25 g |
| Synonym | Hexanedioic-d8 acid (9 CI, ACI), Adipic-d8 acid (6 CI), Hexanedioic-2,2,3,3,4,4,5,5-d8 acid, Hexanedioic acid-D8, Adipic Acid-d8 |
| InChI Formula | InChI=1 S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 2,2,3,3,4,4,5,5-octadeuteriohexanedioic acid |
| Molecular Weight (g/mol) | 154.1905 |
| Formula Weight | 154.1081 g/mol |
| Show More |