Learn More
Hexamethyl-d18-disilazane, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexamethyldisilazane-D18 |
| CAS | 136068-19-4 |
| Molecular Formula | C6 D18 H N Si2 |
| Quantity | 0.25 g |
| Synonym | Silanamine, 1,1,1-tri(methyl-d3)-N-[tri(methyl-d3)silyl]- (9 CI), Hexamethyl-d18-disilazane, 1,1,1-Tri(methyl-d3)-N-[tri(methyl-d3)silyl]silanamine, Hexamethyldisilazane-d18, HMDS-d18, Hexamethyldisilazane-D18 |
| InChI Formula | InChI=1 S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3 |
| SMILES | [2 H]C([2 H])([2 H])[Si](N[Si](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | [bis(trideuteriomethyl)-[tris(trideuteriomethyl)silylamino]silyl]-trideuteriomethane |
| Molecular Weight (g/mol) | 179.5037 |
| Formula Weight | 179.2186 g/mol |
| Show More |