Learn More
(±)-Glycidyl-d5 Hexadecanoate, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Glycidyl Palmitate-d5 |
| CAS | 1794941-80-2 |
| Molecular Formula | C19 D5 H31 O3 |
| Quantity | 0.25 g |
| Synonym | [Dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate, Glycidyl palmitate-d5, Hexadecanoic acid (2-oxiranyl-1,3,3-d3)methyl-d2 ester, Palmitic acid 2,3-epoxypropyl-1,1,2,3,3-d5 ester, (±)-Glycidyl-d5 palmitate, Glycidyl-d5 hexadecanoate, Glycidyl palmitate D5 (glycidyl-D5), Palmitic acid glycidyl-d5 ester |
| InChI Formula | InChI=1 S/C19H36O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(20)22-17-18-16-21-18/h18H,2-17H2,1H3/i16D2,17D2,18 D |
| SMILES | [2 H]C([2 H])(OC(=O)CCCCCCCCCCCCCCC)C1([2 H])OC1([2 H])[2 H] |
| IUPAC Name | [dideuterio-(2,3,3-trideuteriooxiran-2-yl)methyl] hexadecanoate |
| Molecular Weight (g/mol) | 317.518 |
| Formula Weight | 317.298 g/mol |
| Show More |