missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Bromo-2-methyl-d3-propionate-3,3,3-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-2-methylpropanoic Acid Ethyl Ester-D6 (dimethyl-D6) |
| CAS | 64768-38-3 |
| Molecular Formula | C6 D6 H5 Br O2 |
| Quantity | 0.25 g |
| Synonym | Propanoic-3,3,3-d3 acid, 2-bromo-2-(methyl-d3)-, ethyl ester (9 CI), Ethyl 2-Bromo-2-methyl-d3-propionate-3,3,3-d3, 2-Bromo-2-(methyl-d3)propanoic-3,3,3-d3 acid ethyl ester, Ethyl 2-bromo-2-(methyl-d3)propanoate-3,3,3-d3, 2-Bromo-2-methylpropanoic Acid Ethyl Ester-D6 (dimethyl-D6) |
| InChI Formula | InChI=1 S/C6H11BrO2/c1-4-9-5(8)6(2,3)7/h4H2,1-3H3/i2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Br)(C(=O)OCC)C([2 H])([2 H])[2 H] |
| IUPAC Name | ethyl 2-bromo-3,3,3-trideuterio-2-(trideuteriomethyl)propanoate |
| Molecular Weight (g/mol) | 201.0913 |
| Formula Weight | 200.0319 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction