Learn More
DL-Valine-d8, CDN
Product Code. 30324888
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324888
Brand: CDN CDND16980.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Valine-2,3,4,4,4,4',4',4'-D8
CAS 203784-63-8
Molecular Formula C5 D8 H3 N O2
Quantity 0.25 g
Synonym Valine-2,3,4,4,4,4',4',4'-d8, DL-Valine-d8, 2-Amino-3-(methyl-d3)butanoic-2,3,4,4,4-d5 acid
InChI Formula InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/i1D3,2D3,3 D,4 D
SMILES [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])(N)C(=O)O
IUPAC Name 2-amino-2,3,4,4,4-pentadeuterio-3-(trideuteriomethyl)butanoic acid
Molecular Weight (g/mol) 125.1956
Formula Weight 125.1292 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity