Learn More
DL-Methionine-3,3,4,4-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | (±)-Methionine-d4 |
| CAS | 93709-61-6 |
| Molecular Formula | C5H7D4NO2S |
| Quantity | 0.25 g |
| Synonym | DL-Methionine-3,3,4,4-d4, Methionine-3,3,4,4-d4, Acimetion-d4, Amurex-d4, Banthionine-d4, Cynaron-d4, DL-2-Amino-4-(methylthio)butyric Acid-d4, Dyprin-d4, Lactet-d4, Lobamine-d4, Meonine-d4, Meprom M 85-d4, MetAMINO-d4, Methilanin-d4, MetiPEARL-d4, Metione-d4, NSC 9241-d4, Neston-d4, Pedameth-d4, Racemethionine-d4, Rhodimet NP 99-d4, Smartamine-d4, Smartamine M-d4, Timet-d4, Urimeth-d4, α-Amino-γ-methylmercaptobutyric Acid-d4 |
| InChI Formula | InChI=1 S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/i2D2,3D2 |
| SMILES | NC(C(O)=O)C([2 H])([2 H])C([2 H])([2 H])SC |
| IUPAC Name | methionine-3,3,4,4-d4 |
| Molecular Weight (g/mol) | 153.24 |
| Formula Weight | 153.08 g/mol |
| Show More |