missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Hexanedioate-2,2,5,5-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Hexanedioic Acid Dimethyl Ester-2,2,5,5-D4 |
| CAS | 76785-05-2 |
| Molecular Formula | C8 D4 H10 O4 |
| Quantity | 0.25 g |
| Synonym | Hexanedioic-2,2,5,5-d4 acid, dimethyl ester (9 CI), 1,6-Dimethyl hexanedioate-2,2,5,5-d4, Dimethyl Hexanedioate-2,2,5,5-d4, Adipic-2,2,5,5-d4 acid dimethyl ester, Hexanedioic Acid Dimethyl Ester-2,2,5,5-D4 |
| InChI Formula | InChI=1 S/C8H14O4/c1-11-7(9)5-3-4-6-8(10)12-2/h3-6H2,1-2H3/i5D2,6D2 |
| SMILES | [2 H]C([2 H])(CCC([2 H])([2 H])C(=O)OC)C(=O)OC |
| IUPAC Name | dimethyl 2,2,5,5-tetradeuteriohexanedioate |
| Molecular Weight (g/mol) | 178.219 |
| Formula Weight | 178.1143 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction