Learn More
Diethyl-2,2,2,2',2',2'-d6-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Diethylamine 2,2,2,2',2',2'-d6 |
| CAS | 51045-01-3 |
| Molecular Formula | C4 2H6 H5 N |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])CNCC([2 H])([2 H])[2 H] |
| IUPAC Name | 2,2,2-trideuterio-N-(2,2,2-trideuterioethyl)ethanamine |
| Molecular Weight (g/mol) | 79.17 |
| Formula Weight | 79.1268 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |