Learn More
Dibenzothiophene-d8, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Dibenzothiophene D8 |
| CAS | 33262-29-2 |
| Molecular Formula | C12 2H8 S |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C12H8S/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(sc3c([2 H])c([2 H])c([2 H])c([2 H])c23)c1[2 H] |
| IUPAC Name | 1,2,3,4,6,7,8,9-octadeuteriodibenzothiophene |
| Molecular Weight (g/mol) | 192.31 |
| Formula Weight | 192.0849 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |