Learn More
Di-iso-propyl-d14-amine, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N,N-Diisopropylamine-d14 |
| CAS | 70237-41-1 |
| Molecular Formula | C6 D14 H N |
| Quantity | 0.25 g |
| Synonym | 2-Propan-1,1,1,2,3,3,3-d7-amine, N-[1-(methyl-d3)ethyl-1,2,2,2-d4]- (9 CI), N-[1-(Methyl-d3)ethyl-1,2,2,2-d4]-2-propan-1,1,1,2,3,3,3-d7-amine, Di-iso-propyl-d14-amine, Diisopropylamine-d14, Diisopropylamine-D14, N,N-Diisopropylamine-d14 |
| InChI Formula | InChI=1 S/C6H15N/c1-5(2)7-6(3)4/h5-7 H,1-4H3/i1D3,2D3,3D3,4D3,5 D,6 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,3-heptadeuterio-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)propan-2-amine |
| Molecular Weight (g/mol) | 115.2763 |
| Formula Weight | 115.2083 g/mol |
| Show More |