missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl-2,2,3,3-d4-methyl-d2 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Cyclopropylmethanol-1,1,2',2',3',3'-D6 |
| CAS | 1219805-67-0 |
| Molecular Formula | C4 D6 H2 O |
| Quantity | 0.25 g |
| Synonym | Cyclopropyl-2,2,3,3-d4-methyl-d2 Alcohol, Cyclopropane-2,2,3,3-d4-methan-d2-ol, Cyclopropylmethanol-d6, Cyclopropylmethanol-D6 (cyclopropyl-2,2,3,3-D4,methanol-D2), Cyclopropylmethanol-1,1,2',2',3',3'-D6 |
| InChI Formula | InChI=1 S/C4H8O/c5-3-4-1-2-4/h4-5 H,1-3H2/i1D2,2D2,3D2 |
| SMILES | [2 H]C([2 H])(O)C1C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | dideuterio-(2,2,3,3-tetradeuteriocyclopropyl)methanol |
| Molecular Weight (g/mol) | 78.1427 |
| Formula Weight | 78.0952 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction