Learn More
Cyclohexanecarbonyl-d11 Chloride, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Cyclohexanecarbonyl Chloride-D11 |
| CAS | 2708286-89-7 |
| Molecular Formula | C7 D11 Cl O |
| Quantity | 0.25 g |
| Synonym | Cyclohexane-d11-carbonyl chloride, Cyclohexanecarbonyl-d11 Chloride, (Chlorocarbonyl)cyclohexane-d11, Cyclohexanecarboxylic acid chloride-d11, Cyclohexanoic acid chloride-d11, Cyclohexanecarbonyl Chloride-D11 |
| InChI Formula | InChI=1 S/C7H11ClO/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)Cl)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexane-1-carbonyl chloride |
| Molecular Weight (g/mol) | 157.6824 |
| Formula Weight | 157.1189 g/mol |
| Show More |