Learn More
(Bromomethyl)cyclopropane-2,2,3,3-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Cyclopropylmethyl Bromide-2',2',3',3'-D4 |
| CAS | 1219805-93-2 |
| Molecular Formula | C4 D4 H3 Br |
| Quantity | 0.25 g |
| Synonym | 1-(Bromomethyl)cyclopropane-2,2,3,3-d4, 1-(Bromomethyl)-2,2,3,3-tetradeuteriocyclopropane, Cyclopropylmethyl Bromide-d4, (Cyclopropyl-2,2,3,3-d4)methyl Bromide, Cyclopropylmethyl Bromide-2',2',3',3'-D4 |
| InChI Formula | InChI=1 S/C4H7Br/c5-3-4-1-2-4/h4H,1-3H2/i1D2,2D2 |
| SMILES | [2 H]C1([2 H])C(CBr)C1([2 H])[2 H] |
| IUPAC Name | 3-(bromomethyl)-1,1,2,2-tetradeuteriocyclopropane |
| Molecular Weight (g/mol) | 139.027 |
| Formula Weight | 137.9982 g/mol |
| Show More |