Learn More
Bromocyclopropane-2,2,3,3-d4, CDN
Product Code. 30314800
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314800
Brand: CDN CDND74800.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bromocyclopropane-d4
CAS 1398065-55-8
Molecular Formula C3 D4 H Br
Quantity 0.25 g
Synonym Cyclopropyl bromide-d4, NSC 89692-d4
InChI Formula InChI=1 S/C3H5Br/c4-3-1-2-3/h3H,1-2H2/i1D2,2D2
SMILES [2 H]C1([2 H])C(Br)C1([2 H])[2 H]
IUPAC Name 3-bromo-1,1,2,2-tetradeuteriocyclopropane
Molecular Weight (g/mol) 125
Formula Weight 123.9826 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity