Learn More
Benzyl-d7-amine, CDN
Product Code. 30320311
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320311
Brand: CDN CDND52250.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzylamine-d7
CAS 167750-79-0
Molecular Formula C7 D7 H2 N
Quantity 0.25 g
Synonym Benzene-d5-methan-d2-amine (9 CI), Benzyl-d7-amine, Benzylamine-d7, Benzylamine-D7
InChI Formula InChI=1 S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5 H,6,8H2/i1D,2 D,3 D,4 D,5 D,6D2
SMILES [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])N
IUPAC Name dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanamine
Molecular Weight (g/mol) 114.196
Formula Weight 114.117 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity