Learn More
Adamantane-d15-carboxylic Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Adamantane-d15-carboxylic Acid |
| CAS | 33830-12-5 |
| Molecular Formula | C11 D15 H O2 |
| Quantity | 0.25 g |
| Synonym | Tricyclo[3.3.1.13,7 ]decane-2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-d15-1-carboxylic acid (ACI), 1-Adamantane-d15-carboxylic acid (8 CI), 1-Adamantanecarboxylic acid-d15, 1-Adamantyl-d15-carboxylic acid, 1-Carboxyadamantane-d15, Adamantoic acid-d15, Adamantoic Acid-D15 |
| InChI Formula | InChI=1 S/C11H16O2/c12-10(13)11-4-7-1-8(5-11)3-9(2-7)6-11/h7-9 H,1-6H2,(H,12,13)/i1D2,2D2,3D2,4D2,5D2,6D2,7 D,8 D,9 D |
| SMILES | [2 H]C1([2 H])C2([2 H])C([2 H])([2 H])C3([2 H])C([2 H])([2 H])C1([2 H])C([2 H])([2 H])C(C(=O)O)(C3([2 H])[2 H])C2([2 H])[2 H] |
| IUPAC Name | 2,2,3,4,4,5,6,6,7,8,8,9,9,10,10-pentadecadeuterioadamantane-1-carboxylic acid |
| Molecular Weight (g/mol) | 195.336 |
| Formula Weight | 195.2092 g/mol |
| Show More |