missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromopentanoic-3,3,4,4-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 5-Bromopentanoic-3,3,4,4-d4 Acid |
| CAS | 42932-62-7 |
| Molecular Formula | C5 D4 H5 Br O2 |
| Quantity | 0.25 g |
| Synonym | 5-Bromovaleric Acid-d4, 1-Bromo-4-carboxybutane-d4, 5-Bromo-1-pentanoic Acid-d4, 5-Bromopentanoic Acid-d4, 5-Bromovaleric Acid-d4, NSC 53507-d4, delta-Bromovaleric Acid-d4 |
| InChI Formula | InChI=1 S/C5H9BrO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(CBr)C([2 H])([2 H])CC(=O)O |
| IUPAC Name | 5-bromo-3,3,4,4-tetradeuteriopentanoic acid |
| Molecular Weight (g/mol) | 185.05 |
| Formula Weight | 184.0037 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction