missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzoic-2,3,5,6-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Methoxybenzoic Acid D4 (phenyl-D4) |
| CAS | 152404-46-1 |
| Molecular Formula | C8 D4 H4 O3 |
| Quantity | 0.25 g |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-methoxy- (9 CI), 4-Methoxybenzoic-2,3,5,6-d4 acid, 4-Methoxy-2,3,5,6-tetradeuteriobenzoic acid, 4-Methoxybenzoic acid-d4, 4-Methoxybenzoic-d4 acid, 4-Methoxybenzoic acid D4 (phenyl-D4), p-Anisic acid-d4 |
| InChI Formula | InChI=1 S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5 H,1H3,(H,9,10)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1OC |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-methoxybenzoic acid |
| Molecular Weight (g/mol) | 156.172 |
| Formula Weight | 156.072 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction