Learn More
4-Hydroxypiperidine-2,2,3,3,4,5,5,6,6-d9, CDN
Product Code. 30316924
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316924
Brand: CDN CDND65500.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Hydroxypiperidine-d9
CAS 1219799-39-9
Molecular Formula C5H2D9NO
Quantity 0.25 g
Synonym 4-Hydroxypiperidine-2,2,3,3,4,5,5,6,6-d9, 4-Piperidin-2,2,3,3,4,5,5,6,6-d9-ol (ACI)
InChI Formula InChI=1 S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7 H,1-4H2/i1D2,2D2,3 D,4D2,5 D/hD
SMILES OC1([2 H])C([2 H])([2 H])C([2 H])([2 H])N([2 H])C([2 H])C1([2 H])[2 H]
IUPAC Name piperidin-1,2,2,3,3,4,5,5,6-d9-4-ol
Molecular Weight (g/mol) 110.2
Formula Weight 110.14 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity