missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl-2,3,5,6-d4 Chloride, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorobenzylchloride-2,3,5,6-D4 |
| CAS | 1219802-84-2 |
| Molecular Formula | C7 D4 H2 Cl2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,4,5-d4, 3-chloro-6-(chloromethyl)- (ACI), 3-Chloro-6-(chloromethyl)benzene-1,2,4,5-d4 (ACI), 4-Chlorobenzyl-2,3,5,6-d4 Chloride, 4-Chlorobenzylchloride-d4, 4-Chlorobenzylchloride-2,3,5,6-D4, 4-Chloro-1-(chloromethyl)benzene-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H6Cl2/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(CCl)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 1-chloro-4-(chloromethyl)-2,3,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 165.0532 |
| Formula Weight | 164.0098 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction