Learn More
3-Chloroaniline-2,4,6-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-2,4,6-trideuteroaniline |
| CAS | 347840-11-3 |
| Molecular Formula | C6 D3 H3 Cl N |
| Quantity | 0.25 g |
| Synonym | 3-Chlorobenzenamine-d3, 3-Chlorobenzen-2,4,6-d3-amine, 3-Chloroaniline-2,4,6-D3, 3-Chlorobenzen-2,4,6-d3-amine (ACI), 3-Chloro-2,4,6-trideuterioaniline, 3-Chloro-2,4,6-trideuteroaniline, 3-Chloroaniline D3 (2,4,6-phenyl D3) |
| InChI Formula | InChI=1 S/C6H6ClN/c7-5-2-1-3-6(8)4-5/h1-4 H,8H2/i2D,3 D,4 D |
| SMILES | [2 H]c1cc([2 H])c(Cl)c([2 H])c1N |
| IUPAC Name | 3-chloro-2,4,6-trideuterioaniline |
| Molecular Weight (g/mol) | 130.59 |
| Formula Weight | 130.0377 g/mol |
| Show More |