Learn More
3-Bromopropionic-2,2,3,3-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 3-Bromopropanoic Acid-D4 |
| CAS | 1219799-25-3 |
| Molecular Formula | C3 D4 H Br O2 |
| Quantity | 0.25 g |
| Synonym | Propanoic-2,2,3,3-d4 acid, 3-bromo- (ACI), 3-Bromopropanoic-2,2,3,3-d4 acid (ACI), 3-Bromopropionic-2,2,3,3-d4 Acid, 3-Bromopropanoic acid-2,2,3,3-d4, 3-Bromopropanoic Acid-D4 |
| InChI Formula | InChI=1 S/C3H5BrO2/c4-2-1-3(5)6/h1-2H2,(H,5,6)/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(Br)C([2 H])([2 H])C(=O)O |
| IUPAC Name | 3-bromo-2,2,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 156.9993 |
| Formula Weight | 155.9724 g/mol |
| Show More |