Learn More
3,5-Dimethyl-d6-phenol, CDN
Product Code. 30320692
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320692
Brand: CDN CDND56720.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,5-Dimethylphenol-d6
CAS 133604-75-8
Molecular Formula C8H4D6O
Quantity 0.25 g
Synonym NSC 9268-d6, 1-Hydroxy-3,5-dimethylbenzene-d6, 3,5-Xylenol-d6, 1,3,5-Xylenol-d6
InChI Formula InChI=1 S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9 H,1-2H3/i1D3,2D3
SMILES OC1=CC(C([2 H])([2 H])[2 H])=CC(C([2 H])([2 H])[2 H])=C1
IUPAC Name 3,5-bis(methyl-d3)phenol
Molecular Weight (g/mol) 128.2
Formula Weight 128.11 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity