missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylethan-1,1,2,2-d4-ol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Phenylethanol-1,1,2,2-D4 |
| CAS | 107473-33-6 |
| Molecular Formula | C8 D4 H6 O |
| Quantity | 0.25 g |
| Synonym | Benzeneethan-α,α,β,β-d4-ol (9 CI, ACI), Benzeneethan-d4-ol, 2-Phenylethan-1,1,2,2-d4-ol, 2-Phenyl-1,1,2,2-tetradeuterioethanol, 2-Phenylethanol-1,1,2,2-d4, Phenethyl-α,α,β,β-d4 Alcohol, Phenethyl alcohol-d4, 2-Phenylethanol-1,1,2,2-D4 |
| InChI Formula | InChI=1 S/C8H10O/c9-7-6-8-4-2-1-3-5-8/h1-5,9 H,6-7H2/i6D2,7D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])c1ccccc1 |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-phenylethanol |
| Molecular Weight (g/mol) | 126.1891 |
| Formula Weight | 126.0983 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction