Learn More
2-Phenoxyethyl-1,1,2,2-d4 Alcohol, CDN
Product Code. 30310322
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30310322
Brand: CDN CDND56050.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Phenoxy)ethanol-1,1,2,2-D4
CAS 1219804-65-5
Molecular Formula C8H6D4O2
Quantity 0.25 g
Synonym 2-Phenoxyethyl-1,1,2,2-d4 Alcohol
InChI Formula InChI=1 S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9 H,6-7H2/i6D2,7D2
SMILES OC([2 H])(C([2 H])(OC1=CC=CC=C1)[2 H])[2 H]
IUPAC Name 2-phenoxyethan-1,1,2,2-d4-1-ol
Molecular Weight (g/mol) 142.19
Formula Weight 142.09 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity