Learn More
2-Methylpropyl-d9-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Methylpropyl-d9-amine |
| CAS | 1146967-64-7 |
| Molecular Formula | C4 2H9 H2 N |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C4H11N/c1-4(2)3-5/h4H,3,5H2,1-2H3/i1D3,2D3,3D2,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-1-amine |
| Molecular Weight (g/mol) | 82.19 |
| Formula Weight | 82.1456 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |