Learn More
2-Mercapto-5-methoxybenzimidazole-4,6,7-d3, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 5-Methoxy-1 H-benzimidazole-2-thiol-4,6,7-D3 |
| CAS | 1219804-80-4 |
| Molecular Formula | C8 D3 H5 N2 O S |
| Quantity | 0.25 g |
| Synonym | 1,3-Dihydro-5-methoxy-2 H-benzimidazole-2-thione-4,6,7-d3, 5-Methoxy-1 H-benzimidazole-2-thiol-4,6,7-D3, 5-Methoxy-1 H-benzimidazole-2-thiol-d3, 2-Mercapto-5-methoxy-4,6,7-trideuteriobenzimidazole |
| InChI Formula | InChI=1 S/C8H8N2OS/c1-11-5-2-3-6-7(4-5)10-8(12)9-6/h2-4 H,1H3,(H2,9,10,12)/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c2[nH]cnc2c([2 H])c1OC |
| IUPAC Name | 4,6,7-trideuterio-5-methoxy-1 H-benzimidazole-2-thiol |
| Molecular Weight (g/mol) | 183.2454 |
| Formula Weight | 183.0546 g/mol |
| Show More |