missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroacetamide-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Chloroacetamide N,N,2,2-d4 |
| CAS | 122775-20-6 |
| Molecular Formula | C2 2H4 Cl N O |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)/i1D2/hD2 |
| SMILES | [2 H]N([2 H])C(=O)C([2 H])([2 H])Cl |
| IUPAC Name | 2-chloro-N,N,2,2-tetradeuterioacetamide |
| Molecular Weight (g/mol) | 97.54 |
| Formula Weight | 97.0232 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction