missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butoxyethanol-1,1,2,2-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Butoxyethanol (1,1,2,2-D4) |
| CAS | 1219803-96-9 |
| Molecular Formula | C6 D4 H10 O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3/i4D2,6D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])OCCCC |
| IUPAC Name | 2-butoxy-1,1,2,2-tetradeuterio-ethanol |
| Molecular Weight (g/mol) | 122.2 |
| Formula Weight | 122.1245 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction