Learn More
2-Bromobenzoic-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Bromobenzoic Acid-D4 |
| CAS | 2133385-73-4 |
| Molecular Formula | C7 D4 H Br O2 |
| Quantity | 0.25 g |
| Synonym | Benzoic-2,3,4,5-d4 acid, 6-bromo- (ACI), 6-Bromobenzoic-2,3,4,5-d4 acid (ACI), 2-Bromobenzoic-3,4,5,6-d4 acid, 2-Bromobenzoic acid-d4, 2-Bromobenzoic-d4 acid, 2-Bromobenzoic acid-D4 |
| InChI Formula | InChI=1 S/C7H5BrO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(Br)c1[2 H] |
| IUPAC Name | 2-bromo-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 205.0421 |
| Formula Weight | 203.9724 g/mol |
| Show More |