Learn More
2,6-Dimethyl-d6-aniline, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,6-Dimethylaniline-d6 (o-Xylidine-d6) |
| CAS | 919785-81-2 |
| Molecular Formula | C8 2H6 H5 N |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5 H,9H2,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cccc(c1N)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,6-bis(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 127.22 |
| Formula Weight | 127.1268 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |